Imprimer

Anti-PTGS2 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302755EA 454 EUR
ORIGTA302755
Anti-PTGS2 Goat Polyclonal Antibody
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: PTGS2 (prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR