Anti-CBL Goat polyclonal antibody

Supplier: OriGene
ORIGTA302756EA 438 EUR
ORIGTA302756
Anti-CBL Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: CBL (Cbl proto-oncogene, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR