Imprimer

Anti-DUSP8 Goat polyclonal antibody

Fournisseur: OriGene
ORIGTA302764EA 438 EUR InStock
ORIGTA302764
Anti-DUSP8 Goat polyclonal antibody
Anticorps
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: DUSP8 (dual specificity phosphatase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR