Anti-PINX1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302765EA 438 EUR
ORIGTA302765
Anti-PINX1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: PINX1 (PIN2/TERF1 interacting, telomerase inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR