Anti-VAV2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302768EA 502 EUR
ORIGTA302768
Anti-VAV2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-15µg/ml

Type: Primary
Antigen: VAV2 (vav 2 guanine nucleotide exchange factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR