Anti-TMPRSS5 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302769EA 438 EUR
ORIGTA302769
Anti-TMPRSS5 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000 Western Blot: 05-15µg/ml

Type: Primary
Antigen: TMPRSS5 (transmembrane protease, serine 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR