Imprimer

Anti-UBE2V2 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302785EA 547 EUR
ORIGTA302785
Anti-UBE2V2 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 02-1µg/ml

Type: Primary
Antigen: UBE2V2 (ubiquitin-conjugating enzyme E2 variant 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR