Anti-CST7 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302791EA 435 EUR InStock
ORIGTA302791
Anti-CST7 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: CST7 (cystatin F (leukocystatin))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR