Anti-GULP1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302797EA 438 EUR
ORIGTA302797
Anti-GULP1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 02-06µg/ml; Immunochemistry: 5-10 µg/ml

Type: Primary
Antigen: GULP1 (GULP, engulfment adaptor PTB domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR