Anti-KIF5B Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302804EA 438 EUR
ORIGTA302804
Anti-KIF5B Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 02-1µg/ml; Immunochemistry: 1-2µg/ml

Type: Primary
Antigen: KIF5B (kinesin family member 5B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR