Imprimer

Anti-TXNRD1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302806EA 547 EUR
ORIGTA302806
Anti-TXNRD1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-1µg/ml

Type: Primary
Antigen: TXNRD1 (thioredoxin reductase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR