Anti-CBR1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302807EA 500 EUR
ORIGTA302807
Anti-CBR1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000 Western Blot: 02-05µg/ml; Immunochemistry: 1-2µg/ml

Type: Primary
Antigen: CBR1 (carbonyl reductase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR