Anti-SF3B4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302812EA 438 EUR
ORIGTA302812
Anti-SF3B4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: SF3B4 (splicing factor 3b, subunit 4, 49kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR