Anti-ARL4A Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302814EA 438 EUR InStock
ORIGTA302814
Anti-ARL4A Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: ARL4A (ADP-ribosylation factor-like 4A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR