Anti-PAX5 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302816EA 500 EUR
ORIGTA302816
Anti-PAX5 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2-4µg/ml

Type: Primary
Antigen: PAX5 (paired box 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR