Anti-NOS1AP Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302819EA 500 EUR
ORIGTA302819
Anti-NOS1AP Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 5µg/ml

Type: Primary
Antigen: NOS1AP (nitric oxide synthase 1 (neuronal) adaptor protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR