Anti-FOXG1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302825EA 438 EUR InStock
ORIGTA302825
Anti-FOXG1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: FOXG1 (forkhead box G1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR