Anti-POU2AF1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302834EA 438 EUR
ORIGTA302834
Anti-POU2AF1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: POU2AF1 (POU class 2 associating factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR