Anti-RAD51C Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302837EA 500 EUR
ORIGTA302837
Anti-RAD51C Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: RAD51C (RAD51 paralog C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR