Anti-PRDM11 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302841EA 502 EUR
ORIGTA302841
Anti-PRDM11 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 2-3µg/ml

Type: Primary
Antigen: PRDM11 (PR domain containing 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR