Imprimer

Anti-EZH1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302843EA 550 EUR
ORIGTA302843
Anti-EZH1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: EZH1 (enhancer of zeste homolog 1 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR