Anti-UBE2K Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302847EA 500 EUR
ORIGTA302847
Anti-UBE2K Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3 µg/ml

Type: Primary
Antigen: UBE2K (ubiquitin-conjugating enzyme E2K)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR