Anti-CHFR Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302851EA 438 EUR
ORIGTA302851
Anti-CHFR Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000 Western Blot: 25µg/ml

Type: Primary
Antigen: CHFR (checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR