Imprimer

Anti-PCGF3 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302854EA 547 EUR
ORIGTA302854
Anti-PCGF3 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:2000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: PCGF3 (polycomb group ring finger 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR