Anti-NIF3L1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302856EA 438 EUR
ORIGTA302856
Anti-NIF3L1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 02-1µg/ml

Type: Primary
Antigen: NIF3L1 (NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR