Anti-CLPP Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302857EA 438 EUR InStock
ORIGTA302857
Anti-CLPP Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml; Immunochemistry: 2-4?g/ml; ELISA: 1:1000

Type: Primary
Antigen: CLPP (caseinolytic mitochondrial matrix peptidase proteolytic subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR