Anti-TRIM5 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302865EA 438 EUR InStock
ORIGTA302865
Anti-TRIM5 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml; Immunochemistry: 10µg/ml

Type: Primary
Antigen: TRIM5 (tripartite motif containing 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR