Anti-PTBP1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302892EA 438 EUR InStock
ORIGTA302892
Anti-PTBP1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: PTBP1 (polypyrimidine tract binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR