Anti-KALRN Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302900EA 438 EUR
ORIGTA302900
Anti-KALRN Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: KALRN (kalirin, RhoGEF kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR