Anti-PDLIM1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302914EA 438 EUR
ORIGTA302914
Anti-PDLIM1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-1µg/ml

Type: Primary
Antigen: PDLIM1 (PDZ and LIM domain 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR