Anti-EIF4ENIF1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302915EA 438 EUR
ORIGTA302915
Anti-EIF4ENIF1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 025-05µg/ml; Immunochemistry: 5-10µg/ml

Type: Primary
Antigen: EIF4ENIF1 (eukaryotic translation initiation factor 4E nuclear import factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR