Imprimer

Anti-KPNA2 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302917EA 547 EUR
ORIGTA302917
Anti-KPNA2 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000. Western Blot: 0.1-0.5µg/ml. Immunofluorescence: 1:8000

Type: Primary
Antigen: KPNA2 (karyopherin alpha 2 (RAG cohort 1, importin alpha 1))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR