Imprimer

Anti-XPO7 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302918EA 547 EUR
ORIGTA302918
Anti-XPO7 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000. Western Blot: 0.5-1µg/ml

Type: Primary
Antigen: XPO7 (exportin 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR