Anti-VPS29 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302920EA 500 EUR
ORIGTA302920
Anti-VPS29 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: VPS29 (vacuolar protein sorting 29 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR