Imprimer

Anti-STK35 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302922EA 547 EUR
ORIGTA302922
Anti-STK35 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml; Immunochemistry: 10µg/ml

Type: Primary
Antigen: STK35 (serine/threonine kinase 35)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR