Anti-NOXO1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302927EA 438 EUR
ORIGTA302927
Anti-NOXO1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000 Western Blot: 10-20µg/ml

Type: Primary
Antigen: NOXO1 (NADPH oxidase organizer 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR