Imprimer

Anti-ACTR1B Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302940EA 547 EUR
ORIGTA302940
Anti-ACTR1B Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:128000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: ACTR1B (ARP1 actin-related protein 1 homolog B, centractin beta (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR