Anti-CORO1C Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302942EA 502 EUR
ORIGTA302942
Anti-CORO1C Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 02-1µg/ml

Type: Primary
Antigen: CORO1C (coronin, actin binding protein, 1C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR