Anti-LCP2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302947EA 500 EUR
ORIGTA302947
Anti-LCP2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: LCP2 (lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR