Anti-AR Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302952EA 454 EUR
ORIGTA302952
Anti-AR Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-2?g/ml; Immunochemistry: 2-3?g/ml

Type: Primary
Antigen: AR (androgen receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR