Imprimer

Anti-SCARF1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302961EA 547 EUR
ORIGTA302961
Anti-SCARF1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: SCARF1 (scavenger receptor class F, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR