Imprimer

Anti-FNBP1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302962EA 550 EUR
ORIGTA302962
Anti-FNBP1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 025-2µg/ml

Type: Primary
Antigen: FNBP1 (formin binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR