Anti-PTPRT Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302963EA 438 EUR
ORIGTA302963
Anti-PTPRT Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: PTPRT (protein tyrosine phosphatase, receptor type, T)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR