Anti-TIMM50 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302964EA 438 EUR InStock
ORIGTA302964
Anti-TIMM50 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000 Western Blot: 05-3µg/ml

Type: Primary
Antigen: TIMM50 (translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR