Anti-PRDM4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302968EA 502 EUR
ORIGTA302968
Anti-PRDM4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-2µg/ml

Type: Primary
Antigen: PRDM4 (PR domain containing 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR