Imprimer

Anti-FLVCR1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302976EA 547 EUR
ORIGTA302976
Anti-FLVCR1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: FLVCR1 (feline leukemia virus subgroup C cellular receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR