Anti-PSMD2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302978EA 500 EUR
ORIGTA302978
Anti-PSMD2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 003-03µg/ml

Type: Primary
Antigen: PSMD2 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR