Anti-SYNGR4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302981EA 438 EUR
ORIGTA302981
Anti-SYNGR4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 001-01µg/ml

Type: Primary
Antigen: SYNGR4 (synaptogyrin 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR