Anti-MAPK1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302982EA 438 EUR
ORIGTA302982
Anti-MAPK1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: MAPK1 (mitogen-activated protein kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR