Imprimer

Anti-ADH1A, B, C Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302985EA 550 EUR
ORIGTA302985
Anti-ADH1A, B, C Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-15µg/ml

Type: Primary
Antigen: ADH1A, B, C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR